ENA112933327 Enamine

3,3-diethyloxolane-2,5-dione

  • MDL number MFCD08691336
  • Cas Number : 2840-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26420
Availability details for
Quantity
  • Availability
  • Add to Cart