ENA112933330 Enamine

2-(pyridine-3-sulfonamido)acetic acid

  • MDL number MFCD08691338
  • Cas Number : 123090-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26423
Availability details for
Quantity
  • Availability
  • Add to Cart