ENA112933332 Enamine

6-(acetylamino)pyridine-2-carboxylic acid

  • MDL number MFCD00233707
  • Cas Number : 26893-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 226...228
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26425

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26425
Availability details for
Quantity
  • Availability
  • Add to Cart