ENA112933339 Enamine

2-(4-chlorophenyl)-N-methyl-2-(methylamino)acetamide

  • MDL number MFCD08691343
  • Cas Number : 1025448-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26434
Availability details for
Quantity
  • Availability
  • Add to Cart