ENA112933340 Enamine

8-(aminocarbonyl)-2-(2-furyl)imidazo[1,5-a]pyrimidine-4-carboxylic acid

  • MDL number MFCD08691344
  • Cas Number : 924047-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26435

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26435
Availability details for
Quantity
  • Availability
  • Add to Cart