ENA112933341 Enamine

4-{4-imino-5,6-dimethyl-1H,4H-thieno[2,3-d]pyrimidin-2-yl}benzoic acid

  • MDL number MFCD26143699
  • Cas Number : 924209-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26439
Availability details for
Quantity
  • Availability
  • Add to Cart