ENA112933360 Enamine

6-benzyl-5-oxothiomorpholine-3-carboxylic acid

  • MDL number MFCD08691353
  • Cas Number : 31560-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26462

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26462
Availability details for
Quantity
  • Availability
  • Add to Cart