ENA112933362 Enamine

2-[(1,3-benzothiazol-2-yl)methyl]-2-ethylbutanoic acid

  • MDL number MFCD08691354
  • Cas Number : 923716-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26465
Availability details for
Quantity
  • Availability
  • Add to Cart