ENA112933377 Enamine

1-methyl-2-oxo-2,3-dihydro-1H-indole-5-sulfonyl chloride

  • MDL number MFCD08691362
  • Cas Number : 166883-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-158
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26505
Availability details for
Quantity
  • Availability
  • Add to Cart