ENA112933386 Enamine

2-(1-phenyl-1H-pyrazol-4-yl)acetonitrile

  • MDL number MFCD01995560
  • Cas Number : 51412-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26525
Availability details for
Quantity
  • Availability
  • Add to Cart