ENA112933392 Enamine

2-({[(4-fluorophenyl)methyl]carbamoyl}amino)acetic acid

  • MDL number MFCD08691369
  • Cas Number : 923215-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26537

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26537
Availability details for
Quantity
  • Availability
  • Add to Cart