ENA112933394 Enamine

2-({[(4-methylphenyl)methyl]carbamoyl}amino)acetic acid

  • MDL number MFCD08691370
  • Cas Number : 923233-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-26539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26539
Availability details for
Quantity
  • Availability
  • Add to Cart