ENA112933397 Enamine

2-(aminomethyl)-N,N-dimethylbenzene-1-sulfonamide

  • MDL number MFCD08691372
  • Cas Number : 168971-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26543
Availability details for
Quantity
  • Availability
  • Add to Cart