ENA112933398 Enamine

6-(piperidin-1-yl)pyridin-3-amine

  • MDL number MFCD06447399
  • Cas Number : 55403-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26544
Availability details for
Quantity
  • Availability
  • Add to Cart