ENA112933399 Enamine

2-(4-aminopiperidin-1-yl)-N-phenylacetamide

  • MDL number MFCD08691373
  • Cas Number : 923137-83-1
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-26549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26549
Availability details for
Quantity
  • Availability
  • Add to Cart