ENA112933402 Enamine

2-chloro-N-[(isobutylamino)carbonyl]propanamide

  • MDL number MFCD08691376
  • Cas Number : 923106-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26557

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26557
Availability details for
Quantity
  • Availability
  • Add to Cart