ENA112933419 Enamine

2-(2-chloro-N-methylacetamido)-N-[(furan-2-yl)methyl]benzamide

  • MDL number MFCD08691081
  • Cas Number : 923778-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22618

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22618
Availability details for
Quantity
  • Availability
  • Add to Cart