ENA112933429 Enamine

dimethyl-1,3-oxazole-2-thiol

  • MDL number MFCD09040432
  • Cas Number : 6670-14-0
  • Molecular Weight : 129.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4,5-dimethyl-1,3-oxazole-2(3H)-thione
Inchi Code 1S/C5H7NOS/c1-3-4(2)7-5(8)6-3/h1-2H3,(H,6,8)
Inchi Key XEJIHKUDPINBCS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-26856

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-26856
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart