ENA112933433 Enamine

[6-(aminosulfonyl)-2-oxo-1,3-benzoxazol-3(2H)-yl]acetic acid

  • MDL number MFCD09040435
  • Cas Number : 929975-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168...170
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26865

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26865
Availability details for
Quantity
  • Availability
  • Add to Cart