ENA112933435 Enamine

4-[2-(4-phenoxyphenoxy)ethoxy]benzoic acid

  • MDL number MFCD09040436
  • Cas Number : 929975-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26867
Availability details for
Quantity
  • Availability
  • Add to Cart