ENA112933447 Enamine

4-methylpyrimido[1,2-a]benzimidazole-3-carboxylic acid

  • MDL number MFCD03617407
  • Cas Number : 685107-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...201
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26886

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26886
Availability details for
Quantity
  • Availability
  • Add to Cart