ENA112933457 Enamine

2-[4-(piperidin-1-ylsulfonyl)phenyl]isoindolin-1-imine

  • MDL number MFCD03950713
  • Cas Number : 721416-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26900

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26900
Availability details for
Quantity
  • Availability
  • Add to Cart