ENA112933458 Enamine

1-phenyl-4-(prop-2-en-1-yloxy)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD09040449
  • Cas Number : 956264-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26905
Availability details for
Quantity
  • Availability
  • Add to Cart