ENA112933463 Enamine

5-fluoro-2-isocyanatobenzonitrile

  • MDL number MFCD08691395
  • Cas Number : 923232-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26914
Availability details for
Quantity
  • Availability
  • Add to Cart