ENA112933465 Enamine

methyl 6-methyl-2-(methylsulfanyl)pyrimidine-4-carboxylate

  • MDL number MFCD04049012
  • Cas Number : 90007-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-112
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26917

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26917
Availability details for
Quantity
  • Availability
  • Add to Cart