ENA112933469 Enamine

1-isobutyl-4-methyl-1H-pyrazolo[3,4-b]pyridin-6-yl 4-methylbenzenesulfonate

  • MDL number MFCD09040455
  • Cas Number : 929975-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26922

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26922
Availability details for
Quantity
  • Availability
  • Add to Cart