ENA112933472 Enamine

3-(trifluoromethyl)cyclohexanamine

  • MDL number MFCD08444665
  • Cas Number : 56287-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26926

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26926
Availability details for
Quantity
  • Availability
  • Add to Cart