ENA112933481 Enamine

2-(2-chlorophenoxy)aniline

  • MDL number MFCD07365106
  • Cas Number : 56966-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15381
Availability details for
Quantity
  • Availability
  • Add to Cart