ENA112933482 Enamine

5-benzoyl-1,2,3,4-tetrahydropyridine

  • MDL number MFCD02931673
  • Cas Number : 42374-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26937
Availability details for
Quantity
  • Availability
  • Add to Cart