ENA112933483 Enamine

N-(3-{[(2-methoxyphenyl)amino]sulfonyl}-4-methylphenyl)cyclopropanecarboxamide

  • MDL number MFCD08691074
  • Cas Number : 924138-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-15559

Safety Information

msds https://www.enaminestore.com/catalog/EN300-15559
Availability details for
Quantity
  • Availability
  • Add to Cart