ENA112933485 Enamine

3-cyclopropylisoxazol-5-amine

  • MDL number MFCD08558463
  • Cas Number : 21080-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52...54
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26941

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26941
Availability details for
Quantity
  • Availability
  • Add to Cart