ENA112933490 Enamine

6-Methyl-6H-isoindolo[2,1-a]quinazolin-5-one

  • MDL number MFCD00525118
  • Cas Number : 26963-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26950

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26950
Availability details for
Quantity
  • Availability
  • Add to Cart