ENA112933495 Enamine

2-chloro-N-(4-ethyl-2,5-dioxo-4-phenylimidazolidin-1-yl)acetamide

  • MDL number MFCD08691102
  • Cas Number : 923685-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23305

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23305
Availability details for
Quantity
  • Availability
  • Add to Cart