ENA112933507 Enamine

4-[(cyclohexylcarbamoyl)amino]butanoic acid

  • MDL number MFCD08442345
  • Cas Number : 511552-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26982
Availability details for
Quantity
  • Availability
  • Add to Cart