ENA112933508 Enamine

N-[5-(acetylamino)-2-methoxyphenyl]-2-chloroacetamide

  • MDL number MFCD04069123
  • Cas Number : 712287-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23470

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23470
Availability details for
Quantity
  • Availability
  • Add to Cart