ENA112933518 Enamine

5-chloro-2-[(chloroacetyl)amino]benzoic acid

  • MDL number MFCD00157380
  • Cas Number : 14422-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23608

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23608
Availability details for
Quantity
  • Availability
  • Add to Cart