ENA112933521 Enamine

3-(1,3-benzothiazol-2-yl)-4,4,4-trifluoro-3-hydroxybutanenitrile

  • MDL number MFCD06680105
  • Cas Number : 852840-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27035

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27035
Availability details for
Quantity
  • Availability
  • Add to Cart