ENA112933527 Enamine

5-[4-(trifluoromethyl)phenyl]thiophene-2-carbaldehyde

  • MDL number MFCD06410193
  • Cas Number : 343604-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27047

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27047
Availability details for
Quantity
  • Availability
  • Add to Cart