ENA112933531 Enamine

{2-[(methylsulfonyl)amino]-1,3-thiazol-4-yl}acetic acid

  • MDL number MFCD07186383
  • Cas Number : 62557-05-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27056
Availability details for
Quantity
  • Availability
  • Add to Cart