ENA112933535 Enamine

4-(prop-2-en-1-yloxy)-1H-indole-2-carboxylic acid

  • MDL number MFCD08443237
  • Cas Number : 926188-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27061
Availability details for
Quantity
  • Availability
  • Add to Cart