ENA112933539 Enamine

3-(1,3-benzothiazol-2-yl)-4,4,4-trifluoro-3-hydroxybutanoic acid

  • MDL number MFCD08852695
  • Cas Number : 923834-85-9
  • Molecular Weight : 291.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27070
Availability details for
Quantity
  • Availability
  • Add to Cart