ENA112933558 Enamine

11-cyclopropyl-13-phenyl-1,8-diazatricyclo[7.4.0.0,2,7]trideca-2(7),3,5,8,10,12-hexaene-10-carbonitrile

  • MDL number MFCD03976187
  • Cas Number : 732288-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24778

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24778
Availability details for
Quantity
  • Availability
  • Add to Cart