ENA112933574 Enamine

1-(quinazolin-4-yl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD04064789
  • Cas Number : 1008675-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25591
Availability details for
Quantity
  • Availability
  • Add to Cart