ENA113595792 Enamine

1H,2H,5H,6H,7H-cyclopenta[b]pyridin-2-one

  • MDL number MFCD09342899
  • Cas Number : 88499-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-182
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26956
Availability details for
Quantity
  • Availability
  • Add to Cart