ENA113816976 Enamine

2-chloro-N-{[(3-methoxyphenyl)carbamoyl]methyl}-N-methylacetamide

  • MDL number MFCD09040344
  • Cas Number : 929973-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22890

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22890
Availability details for
Quantity
  • Availability
  • Add to Cart