ENA113816988 Enamine

8-benzoyl-6-nitroimidazo[1,2-a]pyridine

  • MDL number MFCD09040505
  • Cas Number : 929972-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-27087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27087
Availability details for
Quantity
  • Availability
  • Add to Cart