ENA113816996 Enamine

3-[5-mercapto-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazol-3-yl]propanamide

  • MDL number MFCD09040513
  • Cas Number : 929972-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27111

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27111
Availability details for
Quantity
  • Availability
  • Add to Cart