ENA113817005 Enamine

5-(thiophen-2-yl)-1-[3-(trifluoromethyl)phenyl]-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD09040522
  • Cas Number : 929972-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27125
Availability details for
Quantity
  • Availability
  • Add to Cart