ENA113817013 Enamine

2,4-dioxo-1-propyl-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD09040529
  • Cas Number : 929972-72-5
  • Molecular Weight : 249.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 244-245
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27143

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-27143
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart