ENA113817016 Enamine

5-methyl-1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09040533
  • Cas Number : 926247-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27168

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27168
Availability details for
Quantity
  • Availability
  • Add to Cart